Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291033.1 | NP_001277962.1 | 4 | 1292 | ||
NM_001291034.1 | NP_001277963.1 | 3 | 1367 | |||
NM_001291035.1 | NP_001277964.1 | 3 | 1508 | |||
NM_001291036.1 | NP_001277965.1 | 4 | 1357 | |||
NM_001291037.1 | NP_001277966.1 | 3 | 1606 | |||
NM_011841.2 | NP_035971.1 | 4 | 1242 | |||
GenBank mRNA | AB019373.1 | - | 4 | 1192 | ||
AF126159.1 | - | 3 | 1005 | |||
AF126160.1 | - | 3 | 1055 | |||
AF126161.1 | - | 3 | 1425 | |||
AK148119.1 | - | 4 | 1224 | |||
AK155187.1 | - | 4 | 1160 | |||
AY534740.1 | - | 4 | 1176 | |||
BC013745.1 | - | 3 | 1525 | |||
BC033598.1 | - | 4 | 1332 | |||
BC051020.1 | - | 2 | 1886 | |||
BC058096.1 | - | 2 | 1886 | |||
BC082315.1 | - | 3 | 1429 |