Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166493.1 | NP_001159965.1 | 5 | 707 | ||
NM_207246.4 | NP_997129.1 | 6 | 699 | |||
GenBank mRNA | AB154824.1 | - | 4 | 394 | ||
AK173041.1 | - | 5 | 592 | |||
BC066069.1 | - | 4 | 337 | 6851971 | ||
BC066127.1 | - | 6 | 624 | 30280332 | ||
CD348455.1 | - | 4 | 347 |