Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF037205.1 | - | 2 | 145 | ||
AF037206.1 | - | 2 | 145 | |||
AI882163.1 | - | 2 | 204 | |||
AK034135.1 | - | 2 | 196 | |||
AK036591.1 | - | 2 | 190 | |||
AK050095.1 | - | 2 | 158 | |||
AK082296.1 | - | 2 | 186 | |||
AK139447.1 | - | 2 | 151 | |||
AK140682.1 | - | 2 | 200 | |||
AK158046.1 | - | 2 | 186 | |||
AK167278.1 | - | 2 | 150 | |||
BB847350.1 | - | 2 | 210 | |||
BC058182.1 | - | 2 | 153 | 5324040 |