Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lipk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205349.1 | NP_001192278.1 | 5, 6 | 682 | ||
NM_172837.4 | NP_766425.1 | 5, 6 | 667 | |||
GenBank mRNA | AK037106.1 | - | 5, 6 | 632 | ||
AK162706.1 | - | 5, 6 | 589 | |||
BC037229.1 | - | 6, 7 | 634 | |||
BC055815.1 | - | 5, 6 | 635 | 4011597 |