Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcmdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177722.3 | NP_808390.2 | 9 | 1056 | ||
GenBank mRNA | AK031395.1 | - | 9 | 1014 | ||
CA978963.1 | - | 7 | 811 |