Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172844.2 | NP_766432.1 | 8 | 1241 | ||
GenBank mRNA | AK029219.1 | - | 8 | 1242 | ||
AK161224.1 | - | 8 | 1241 | |||
BC111885.1 | - | 6 | 970 | 40060432 | ||
BC112405.1 | - | 8 | 1231 | 40060428 |