Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290572.1 | NP_001277501.1 | 3, 4 | 463 | ||
NM_175211.5 | NP_780420.1 | 3, 4 | 463 | |||
GenBank mRNA | AK017939.1 | - | 3, 4 | 439 | ||
AK076767.1 | - | 3, 4 | 448 | |||
AK170248.1 | - | 3, 4 | 409 | |||
BC072656.1 | - | 3, 4 | 398 | 30536315 | ||
CJ112478.1 | - | 3, 4 | 465 |