Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165954.1 | NP_001159426.1 | 14 | 2742 | ||
NM_001165955.1 | NP_001159427.1 | 13 | 2658 | |||
NM_001165956.1 | NP_001159428.1 | 15 | 2616 | |||
NM_153421.2 | NP_700470.2 | 14 | 2757 | |||
GenBank mRNA | AJ414610.1 | - | 14 | 2743 | ||
AK037348.1 | - | 15 | 2802 | |||
AK087339.1 | - | 13 | 2653 | |||
AK166208.1 | - | 14 | 2537 | |||
BC044839.1 | - | 7 | 1418 | |||
BC132533.1 | - | 14 | 2726 | 40130810 | ||
BC145123.1 | - | 15 | 2600 | 9053671 | ||
BC145124.1 | - | 15 | 2575 |