Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033350.2 | NP_001028522.2 | 9 | 1458 | ||
GenBank mRNA | AK041242.1 | - | 9 | 1419 | ||
AK156201.1 | - | 9 | 1468 | |||
AY178734.1 | - | 9 | 1478 | |||
BC026932.1 | - | 9 | 1405 | 4008261 | ||
EU051378.1 | - | 8 | 1059 |