Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Napepld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178728.5 | NP_848843.1 | 2 | 222 | ||
GenBank mRNA | AB112350.1 | - | 1 | 65 | ||
AK041194.1 | - | 2 | 220 | |||
AK049135.1 | - | 2 | 481 | |||
AK080888.1 | - | 2 | 222 | |||
AK141585.1 | - | 2 | 502 | |||
BC062890.1 | - | 2 | 244 | 5702354 |