Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uroc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144940.2 | NP_659189.2 | 20 | 1964 | ||
GenBank mRNA | AK149464.1 | - | 20 | 2067 | ||
BC022133.1 | - | 20 | 1947 | 5098655 | ||
BC033269.1 | - | 17 | 1579 |