Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prickle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081146.2 | NP_001074615.1 | 9 | 1942 | ||
NM_001134459.1 | NP_001127931.1 | 8 | 2021 | |||
NM_001134460.2 | NP_001127932.1 | 8 | 2343 | |||
NM_001134461.1 | NP_001127933.1 | 8 | 1838 | |||
GenBank mRNA | AK032631.1 | - | 8 | 2350 | ||
AK147576.1 | - | 8 | 2022 | |||
AK147628.1 | - | 8 | 2022 | |||
BC050793.1 | - | 8 | 1838 | |||
BC099897.1 | - | 9 | 1913 | 30621797 | ||
BC145754.1 | - | 8 | 1860 | 40131057 |