Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prickle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081146.2 | NP_001074615.1 | 9 | 2515 | ||
NM_001134459.1 | NP_001127931.1 | 8 | 2594 | |||
NM_001134460.2 | NP_001127932.1 | 8 | 2916 | |||
NM_001134461.1 | NP_001127933.1 | 8 | 2411 | |||
GenBank mRNA | AK032631.1 | - | 8 | 2923 | ||
AK147576.1 | - | 8 | 2594 | |||
AK147628.1 | - | 8 | 2594 | |||
BC050793.1 | - | 8 | 2411 | |||
BC099897.1 | - | 9 | 2486 | 30621797 | ||
BC145754.1 | - | 8 | 2433 | 40131057 |