Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172152.1 | NP_001165623.1 | 5 | 763 | ||
NM_028220.3 | NP_082496.3 | 6 | 786 | |||
GenBank mRNA | AK013889.1 | - | 5 | 743 | ||
AK020995.1 | - | 6 | 861 | |||
AK044241.1 | - | 5 | 764 | |||
BC028268.1 | - | 6 | 797 | 5366098 |