Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172918.4 | NP_766506.4 | 9 | 2143 | ||
GenBank mRNA | AK030030.1 | - | 11 | 2283 | ||
AK045365.1 | - | 7 | 1866 | |||
AK051946.1 | - | 9 | 2144 | |||
BC059211.1 | - | 9 | 2106 | 6417014 |