Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029868.2 | NP_001025039.1 | 8 | 2768 | ||
NM_001290540.1 | NP_001277469.1 | 6 | 2423 | |||
GenBank mRNA | AK046101.1 | - | 6 | 2428 | ||
BC056462.1 | - | 8 | 2114 | 5703229 |