Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007576.2 | NP_001007577.1 | 9 | 2170 | ||
GenBank mRNA | AK044234.1 | - | 9 | 2171 | ||
AY651761.1 | - | 9 | 2165 | |||
BC115714.1 | - | 8 | 2000 | 40085764 |