Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139310.1 | NP_647471.1 | 22, 23 | 2510 | ||
GenBank mRNA | AK157904.1 | - | 22, 23 | 2542 | ||
AK158280.1 | - | 12, 13 | 1483 | |||
AK158326.1 | - | 12, 13 | 1479 | |||
AY055122.1 | - | 22, 23 | 2510 |