Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr437 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146296.1 | NP_666408.1 | 1 | 18 | ||
GenBank mRNA | BC137777.1 | - | 1 | 68 | 8860796 |