Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153424.2 | NP_700473.2 | 15 | 2256 | ||
GenBank mRNA | AF537131.1 | - | 13 | 2003 | ||
AK038879.1 | - | 15 | 2011 | |||
AK084539.1 | - | 15 | 2258 | |||
AK172998.1 | - | 12 | 1569 | |||
AY118229.1 | - | 14 | 1948 | |||
BC138370.1 | - | 15 | 1981 | 8861390 |