Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029926.5 | NP_084202.2 | 2 | 233 | ||
GenBank mRNA | AF445803.1 | - | 2 | 221 | ||
AK020397.2 | - | 2 | 241 | |||
AK028837.1 | - | 2 | 174 | |||
AK029028.1 | - | 2 | 234 | |||
AK139992.1 | - | 222 | ||||
BC051676.1 | - | 2 | 226 | 30017484 |