Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3pxd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177364.3 | NP_796338.2 | 7 | 620 | ||
GenBank mRNA | AB430861.1 | - | 7 | 542 | ||
AK089330.1 | - | 7 | 592 | |||
AK090119.1 | - | 7 | 613 | |||
AK154898.1 | - | 7 | 578 | |||
AK163471.1 | - | 7 | 592 | |||
AK169978.1 | - | 7 | 612 | |||
AK171384.1 | - | 7 | 592 | |||
BC115711.1 | - | 7 | 589 | |||
BC115764.1 | - | 7 | 589 | |||
BC141306.1 | - | 7 | 592 | 9055918 |