Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172951.3 | NP_766539.2 | 5 | 486 | ||
GenBank mRNA | AF367760.1 | - | 5 | 341 | ||
AK008673.1 | - | 5 | 394 | |||
AK035310.1 | - | 5 | 440 | |||
AK087560.1 | - | 5 | 436 | |||
AK137345.1 | - | 5 | 445 | |||
BC058204.1 | - | 5 | 376 | |||
BC138956.1 | - | 5 | 400 | 8861978 | ||
BC145476.1 | - | 5 | 395 |