Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024645.1 | NP_001019816.1 | 28 | 2660 | ||
GenBank mRNA | AK131164.1 | - | 28 | 2603 | ||
AK148355.1 | - | 28 | 2683 | |||
AK158466.1 | - | 29 | 2906 | |||
BC036313.1 | - | 13 | 1222 | |||
BC060252.1 | - | 9 | 1200 | |||
BC082805.1 | - | 28 | 2580 | |||
BC092223.1 | - | 28 | 2578 |