Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gulo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178747.3 | NP_848862.1 | 9 | 982 | ||
GenBank mRNA | AK077740.1 | - | 9 | 983 | ||
AK167460.1 | - | 9 | 971 | |||
AY453064.1 | - | 8 | 819 | |||
BC019856.1 | - | 9 | 929 | 5123684 | ||
BC028822.1 | - | 9 | 938 | 5097681 | ||
BC028828.1 | - | 9 | 941 | 5101228 | ||
BC034835.1 | - | 8 | 738 |