Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cln8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012000.3 | NP_036130.1 | 2 | 785 | ||
GenBank mRNA | AF125307.1 | - | 2 | 275 | ||
AF125308.1 | - | 2 | 275 | |||
AK049145.1 | - | 2 | 458 | |||
AK080049.1 | - | 2 | 293 | |||
AK088300.1 | - | 2 | 439 | |||
AK089692.1 | - | 2 | 434 | |||
AK128890.1 | - | 2 | 456 | |||
AK164268.1 | - | 2 | 458 | |||
AK166905.1 | - | 2 | 1210 | |||
AK169698.1 | - | 2 | 439 | |||
AK171446.1 | - | 2 | 467 | |||
BC021625.1 | - | 2 | 291 | 4188527 |