Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkmyt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023058.3 | NP_075545.2 | 3 | 816 | ||
GenBank mRNA | AF175892.1 | - | 3 | 700 | ||
AK031787.1 | - | 2 | 1683 | |||
AK158550.1 | - | 3 | 817 | |||
AK165164.1 | - | 3 | 683 | |||
AK181294.1 | - | 2 | 58 | |||
BC025061.1 | - | 3 | 539 | 2609421 |