Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caskin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027937.2 | NP_082213.2 | 6, 7 | 802 | ||
GenBank mRNA | AK014376.1 | - | 6, 7 | 979 | ||
AK129327.1 | - | 6, 7 | 578 | |||
BC060720.1 | - | 6, 7 | 566 | |||
BC138443.1 | - | 6, 7 | 710 | 8861463 | ||
BC145281.1 | - | 6, 7 | 710 | 9053835 |