Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013767.2 | NP_001013789.1 | 6 | 680 | ||
GenBank mRNA | AY578330.1 | - | 6 | 680 |