Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172964.4 | NP_766552.3 | 17 | 2175 | ||
GenBank mRNA | AK087523.1 | - | 17 | 2188 | ||
AK133720.1 | - | 17 | 2014 | |||
AK141901.1 | - | 17 | 2125 | |||
AK162051.1 | - | 11 | 1215 | |||
AK173147.1 | - | 16 | 1802 |