Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199021.3 | NP_950186.3 | 20 | 1878 | ||
GenBank mRNA | AK032579.1 | - | 18 | 1524 | ||
AK046842.1 | - | 20 | 1879 | |||
BC029696.1 | - | 5 | 360 | |||
BC063074.1 | - | 10 | 800 | 5697531 | ||
BC067026.1 | - | 20 | 1871 | 6853947 |