Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012014.3 | NP_036144.2 | 3 | 2094 | ||
GenBank mRNA | AF146569.1 | - | 1 | 1509 | ||
AK138306.1 | - | 3 | 2095 | |||
AK144210.1 | - | 2 | 2004 | |||
BC037191.1 | - | 1 | 390 | |||
BC057044.1 | - | 2 | 1981 | 6825304 |