Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081162.1 | NP_001074631.1 | 13 | 1637 | ||
GenBank mRNA | AK052665.1 | - | 8 | 2506 | ||
AK158115.1 | - | 13 | 1591 | |||
BC036154.1 | - | 3 | 1215 | |||
BC111884.1 | - | 12 | 1432 | 40060299 |