siRNA Details
Gene Symbol: Tox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098799.1 | NP_001092269.1 | 4 | 666 | ||
GenBank mRNA | BF470404.1 | - | 2 | 462 |

Gene Symbol: Tox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098799.1 | NP_001092269.1 | 4 | 666 | ||
GenBank mRNA | BF470404.1 | - | 2 | 462 |