Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177732.1 | NP_001171203.1 | 15 | 1937 | ||
NM_001177733.1 | NP_001171204.1 | 15 | 1937 | |||
NM_183191.3 | NP_899014.2 | 15 | 1937 | |||
GenBank mRNA | AK173095.1 | - | 7 | 820 | ||
AY691172.1 | - | 15 | 1937 | |||
AY691173.1 | - | 15 | 1937 | |||
AY691174.1 | - | 15 | 1937 | |||
BC042549.1 | - | 3 | 266 | |||
BC052372.1 | - | 3 | 266 | |||
BC055005.1 | - | 15 | 2011 | 6491217 |