Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mms22l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199467.2 | NP_955761.2 | 1, 2 | 598 | ||
GenBank mRNA | AK144799.1 | - | 1, 2 | 517 | ||
AK164069.1 | - | 1, 2 | 598 | |||
AK166072.1 | - | 1, 2 | 609 | |||
CK639437.1 | - | 1, 2 | 150 |