Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024142.1 | NP_001019313.1 | 8, 9 | 2303 | ||
GenBank mRNA | AK142389.1 | - | 7, 8 | 2227 | ||
BC052485.1 | - | 7, 8 | 1770 | |||
BC054731.1 | - | 8, 9 | 2293 | |||
BC062126.1 | - | 7, 8 | 1661 | |||
BC079571.1 | - | 2, 3 | 419 | |||
BC094565.1 | - | 7, 8 | 2087 | 6836944 |