Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039169.1 | NP_001034258.1 | 5 | 945 | ||
NM_001039170.1 | NP_001034259.1 | 5 | 494 | |||
NM_023314.3 | NP_075803.2 | 5 | 494 | |||
GenBank mRNA | AF068116.1 | - | 5 | 408 | ||
AK017221.1 | - | 5 | 946 | |||
AK018516.1 | - | 5 | 492 | |||
AK032965.1 | - | 5 | 498 | |||
AK076067.1 | - | 3 | 209 | |||
AK138055.1 | - | 5 | 433 | |||
AK150495.1 | - | 5 | 433 | |||
BC028903.1 | - | 5 | 544 | 4166275 | ||
BC045153.1 | - | 5 | 428 | 6441327 | ||
BC049077.1 | - | 5 | 462 | 5708556 | ||
U01137.1 | - | 5 | 441 |