Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173445.4 | NP_775621.2 | 4 | 1306 | ||
GenBank mRNA | AK030872.1 | - | 4 | 1277 | ||
AK084219.1 | - | 3 | 1144 | |||
AK088482.1 | - | 3 | 1470 | |||
AK088662.1 | - | 4 | 947 | |||
BC069844.1 | - | 4 | 1286 | 6487223 |