Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033371.3 | NP_001028543.1 | 3 | 315 | ||
NM_001170788.1 | NP_001164259.1 | 6 | 638 | |||
NM_001170789.1 | NP_001164260.1 | 3 | 315 | |||
GenBank mRNA | AK133042.1 | - | 3 | 315 | ||
BC150957.1 | - | 3 | 307 | 9087868 | ||
BC172120.1 | - | 3 | 307 | 9054406 |