Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048143.2 | NP_001041608.1 | 5 | 1561 | ||
NM_173015.3 | NP_766603.1 | 4 | 1751 | |||
GenBank mRNA | AK030288.1 | - | 4 | 1717 | ||
AK157390.1 | - | 5 | 1564 | |||
AK173047.1 | - | 4 | 1423 | |||
BC062639.1 | - | 4 | 1427 | |||
BQ831297.1 | - | 2 | 164 |