Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012056.2 | NP_036186.2 | 7 | 1257 | ||
GenBank mRNA | AF090334.1 | - | 7 | 1183 | ||
AF279263.1 | - | 7 | 1235 | |||
AK007499.1 | - | 5 | 647 | |||
AK218619.1 | - | 1 | 30 | |||
BC026133.1 | - | 7 | 1197 | 4018235 | ||
BC043129.1 | - | 7 | 1257 | 6810029 |
Gene Symbol: Cbfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK198845.1 | - | 262 |