Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tstd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173033.3 | NP_766621.3 | 8 | 1183 | ||
GenBank mRNA | AK028258.1 | - | 8 | 1126 | ||
AK155456.1 | - | 8 | 1130 | |||
BC057893.1 | - | 8 | 1118 | 4978941 |