Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013752.3 | NP_038780.3 | 4 | 560 | ||
GenBank mRNA | AB016988.1 | - | 4 | 482 | ||
AF076687.1 | - | 4 | 554 | |||
AF092840.1 | - | 4 | 543 | |||
AK031933.1 | - | 4 | 546 | |||
AK132302.1 | - | 4 | 601 | |||
AK134805.1 | - | 4 | 596 | |||
AK134960.1 | - | 4 | 588 | |||
AK159790.1 | - | 4 | 567 | |||
AK160614.1 | - | 4 | 526 | |||
BC003719.1 | - | 4 | 571 | |||
BC013190.1 | - | 4 | 560 | |||
BC044773.2 | - | 4 | 559 | 5067851 | ||
BC055061.1 | - | 4 | 552 | 6528890 |