Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yme1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013771.5 | NP_038799.1 | 13 | 1548 | ||
GenBank mRNA | AF090430.1 | - | 13 | 1526 | ||
AK045141.1 | - | 13 | 1533 | |||
AK079175.1 | - | 13 | 1508 | |||
AK137665.1 | - | 13 | 1496 | |||
AY136286.1 | - | 13 | 1364 | |||
AY136287.1 | - | 13 | 1517 | |||
BC007128.1 | - | 13 | 1509 | 3591123 |