Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013850.1 | NP_038878.1 | 13 | 1820 | ||
GenBank mRNA | AF287141.1 | - | 13 | 1820 | ||
BC145120.1 | - | 13 | 1859 | 9053668 | ||
BC150708.1 | - | 13 | 1859 | 9087619 |