Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190443.1 | NP_001177372.1 | 12 | 1602 | ||
NM_013853.2 | NP_038881.1 | 12 | 1605 | |||
GenBank mRNA | AF213382.1 | - | 5 | 319 | ||
AK137233.1 | - | 12 | 1537 | |||
AK147937.1 | - | 12 | 1560 | |||
AK151545.1 | - | 12 | 1561 | |||
BC003300.1 | - | 11 | 1497 | 3583837 |