Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077595.2 | NP_001071063.1 | 7 | 4584 | ||
NM_001077596.2 | NP_001071064.1 | 7 | 4632 | |||
NM_015756.2 | NP_056571.2 | 8 | 4956 | |||
GenBank mRNA | AF199421.1 | - | 8 | 4974 | ||
AF199422.1 | - | 7 | 4542 | |||
AK049347.1 | - | 4 | 2490 | |||
AK129371.1 | - | 4 | 3962 | |||
AK147493.1 | - | 7 | 4567 | |||
AK164918.1 | - | 7 | 4543 | |||
BC003909.1 | - | 2 | 228 | |||
BC145113.1 | - | 9 | 4373 | 9053661 |