Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178760.4 | NP_848875.2 | 9 | 883 | ||
GenBank mRNA | AK030433.1 | - | 9 | 879 | ||
AK034815.1 | - | 9 | 866 | |||
AK082237.1 | - | 9 | 881 | |||
AK083025.1 | - | 9 | 877 | |||
AK088544.1 | - | 9 | 867 | |||
AK129407.1 | - | 9 | 778 | |||
AK142454.1 | - | 9 | 848 | |||
AK154530.1 | - | 9 | 879 | |||
BC092231.1 | - | 9 | 851 | 30607603 | ||
BC117900.1 | - | 9 | 861 | 40097045 | ||
BC117901.1 | - | 9 | 861 | 40097052 |
Set Membership: |
G-protein coupled receptor |