Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017985.2 | NP_001017985.2 | 27 | 5360 | ||
GenBank mRNA | AK047412.1 | - | 21 | 4018 | ||
BC046408.1 | - | 7 | 1457 | |||
BC046540.1 | - | 7 | 1457 | |||
BC094430.1 | - | 26 | 4941 | 30643564 |